2-Bromo-3-chloro-6-fluorobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC501782-1G PC501782-5G PC501782-250MG
APOSPC501782-1GEA 708 EUR
APOSPC501782-1G APOSPC501782-5G APOSPC501782-250MG
2-Bromo-3-chloro-6-fluorobenzyl bromide
2-Bromo-3-chloro-6-fluorobenzyl bromide
Formel: C₇H₄Br₂ClF
Molecular Weight: 302,366 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28141933
CAS-Nummer: 1823559-40-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR