2-Bromo-3-chloro-6-fluorobenzaldehyde

Lieferant: Apollo Scientific
Achtung

PC501788-5G
APOSPC501788-5GEA 186 EUR
APOSPC501788-5G
2-Bromo-3-chloro-6-fluorobenzaldehyde
2-Bromo-3-chloro-6-fluorobenzaldehyde
Formel: C₇H₃BrClFO
Molecular Weight: 237,454 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520667
CAS-Nummer: 1056264-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR