5-Bromo-2,3,4-trifluorobenzeneboronic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Bromo-2,3,4-trifluorophenyl)boronic acid, 5-Bromo-2,3,4-trifluorophenylboronic acid
PC50182-1G
PC50182-250MG
PC50182-5G
APOSPC50182-1GEA
522
EUR
APOSPC50182-1G
APOSPC50182-250MG
APOSPC50182-5G
5-Bromo-2,3,4-trifluorobenzeneboronic acid 95%
5-Bromo-2,3,4-trifluorobenzeneboronic acid
Formel:
C₆H₃BBrF₃O₂ Molecular Weight: 254,797 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12032026 CAS-Nummer: 2377611-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...