2-Bromo-3-chloro-6-fluorobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC501833-1G PC501833-5G
APOSPC501833-1GEA 265 EUR
APOSPC501833-1G APOSPC501833-5G
2-Bromo-3-chloro-6-fluorobenzyl alcohol
2-Bromo-3-chloro-6-fluorobenzyl alcohol
Formel: C₇H₅BrClFO
Molecular Weight: 239.469 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28141932
CAS-Nummer: 1823576-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR