2-Bromo-3,4-difluorobenzonitrile 95%

Lieferant: Apollo Scientific
Achtung

PC501846-5G
APOSPC501846-5GEA 183 EUR
APOSPC501846-5G
2-Bromo-3,4-difluorobenzonitrile 95%
2-Bromo-3,4-difluorobenzonitrile
Formel: C₇H₂BrF₂N
Molecular Weight: 217,998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22490573
CAS-Nummer: 1517611-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR