3-(Difluoromethoxy)-4-fluoronitrobenzene

Lieferant: Apollo Scientific
Achtung

PC501862-1G PC501862-250MG PC501862-5G
APOSPC501862-1GEA 248 EUR
APOSPC501862-1G APOSPC501862-250MG APOSPC501862-5G
3-(Difluoromethoxy)-4-fluoronitrobenzene
3-(Difluoromethoxy)-4-fluoronitrobenzene
Formel: C₇H₄F₃NO₃
Molecular Weight: 207,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698450
CAS-Nummer: 97963-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR