3-Bromo-2-(difluoromethoxy)nitrobenzene

Lieferant: Apollo Scientific
Achtung

PC501873-1G PC501873-250MG PC501873-5G
APOSPC501873-1GEA 58 EUR
APOSPC501873-1G APOSPC501873-250MG APOSPC501873-5G
3-Bromo-2-(difluoromethoxy)nitrobenzene
3-Bromo-2-(difluoromethoxy)nitrobenzene
Formel: C₇H₄BrF₂NO₃
Molecular Weight: 268,012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18390951
CAS-Nummer: 259243-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR