5-Bromo-2-(difluoromethoxy)nitrobenzene 97%

Lieferant: Apollo Scientific
Achtung

PC501880-1G PC501880-250MG
APOSPC501880-1GEA 538 EUR
APOSPC501880-1G APOSPC501880-250MG
5-Bromo-2-(difluoromethoxy)nitrobenzene 97%
5-Bromo-2-(difluoromethoxy)nitrobenzene
Formel: C₇H₄BrF₂NO₃
Molecular Weight: 268,012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18390955
CAS-Nummer: 1261552-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR