2-Bromo-3,5-difluorobenzonitrile 97%

Lieferant: Apollo Scientific
Gefahr

PC501883-1G PC501883-250MG PC501883-5G
APOSPC501883-1GEA 290 EUR
APOSPC501883-1G APOSPC501883-250MG APOSPC501883-5G
2-Bromo-3,5-difluorobenzonitrile 97%
2-Bromo-3,5-difluorobenzonitrile
Formel: C₇H₂BrF₂N
Molecular Weight: 217,998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22571442
CAS-Nummer: 425379-37-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR