2-Bromo-3,4-difluorobenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC501887-100MG
PC501887-1G
APOSPC501887-100MGEA
21.9
EUR
APOSPC501887-100MG
APOSPC501887-1G
2-Bromo-3,4-difluorobenzyl bromide 95%
2-Bromo-3,4-difluorobenzyl bromide
Formel:
C₇H₄Br₂F₂ Molecular Weight: 285,911 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28737917 CAS-Nummer: 1805523-88-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...