5-Bromo-2-(trifluoromethyl)pyridine-3-carbonitrile 95%
Lieferant: Apollo Scientific
PC50189-1G
PC50189-250MG
APOSPC50189-1GEA
0
EUR
APOSPC50189-1G
APOSPC50189-250MG
5-Bromo-2-(trifluoromethyl)pyridine-3-carbonitrile 95%
5-Bromo-2-(trifluoromethyl)pyridine-3-carbonitrile
Formel:
C₇H₂BrF₃N₂ Molecular Weight: 251,003 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11848373 CAS-Nummer: 1245914-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...