3-Bromo-5-fluoro-2-iodoanisole 95%

Lieferant: Apollo Scientific
Achtung

PC501894-1G PC501894-250MG
APOSPC501894-1GEA 522 EUR
APOSPC501894-1G APOSPC501894-250MG
3-Bromo-5-fluoro-2-iodoanisole 95%
3-Bromo-5-fluoro-2-iodoanisole
Formel: C₇H₅BrFIO
Molecular Weight: 330,921 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29049290
CAS-Nummer: 1935372-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR