2-(Difluoromethoxy)-5-fluoronitrobenzene

Lieferant: Apollo Scientific
Achtung

PC501897-10G PC501897-1G PC501897-5G
APOSPC501897-10GEA 571 EUR
APOSPC501897-10G APOSPC501897-1G APOSPC501897-5G
2-(Difluoromethoxy)-5-fluoronitrobenzene
2-(Difluoromethoxy)-5-fluoronitrobenzene
Formel: C₇H₄F₃NO₃
Molecular Weight: 207,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698447
CAS-Nummer: 1214349-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR