5-Bromo-2,3-difluorobenzaldehyde 96%

Lieferant: Apollo Scientific
Achtung

PC501901-1G PC501901-5G PC501901-10G PC501901-25G
APOSPC501901-1GEA 21.9 EUR
APOSPC501901-1G APOSPC501901-5G APOSPC501901-10G APOSPC501901-25G
5-Bromo-2,3-difluorobenzaldehyde 96%
5-Bromo-2,3-difluorobenzaldehyde
Formel: C₇H₃BrF₂O
Molecular Weight: 220,999 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08669446
CAS-Nummer: 633327-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR