4-Bromo-2-(difluoromethoxy)nitrobenzene 95%

Lieferant: Apollo Scientific

PC501912-10G PC501912-5G
APOSPC501912-10GEA 2050 EUR
APOSPC501912-10G APOSPC501912-5G
4-Bromo-2-(difluoromethoxy)nitrobenzene 95%
4-Bromo-2-(difluoromethoxy)nitrobenzene
Formel: C₇H₄BrF₂NO₃
Molecular Weight: 268,012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18390954
CAS-Nummer: 1261849-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR