4-Bromo-6-fluoro-1,3-benzoxazole
Lieferant: Apollo Scientific
|
Achtung
|
PC501921-1G
PC501921-250MG
APOSPC501921-1GEA
580
EUR
APOSPC501921-1G
APOSPC501921-250MG
4-Bromo-6-fluoro-1,3-benzoxazole
4-Bromo-6-fluoro-1,3-benzoxazole
Formel:
C₇H₃BrFNO Molecular Weight: 216,007 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29049291 CAS-Nummer: 1929606-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...