5-Bromo-2,3-difluorobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC501930-1G PC501930-250MG PC501930-5G
APOSPC501930-1GEA 567 EUR
APOSPC501930-1G APOSPC501930-250MG APOSPC501930-5G
5-Bromo-2,3-difluorobenzyl bromide
5-Bromo-2,3-difluorobenzyl bromide
Formel: C₇H₄Br₂F₂
Molecular Weight: 285,911 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972475
CAS-Nummer: 887585-85-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR