4-Bromo-5-fluoro-2-hydroxybenzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501940-1G
PC501940-250MG
PC501940-5G
APOSPC501940-1GEA
116
EUR
APOSPC501940-1G
APOSPC501940-250MG
APOSPC501940-5G
4-Bromo-5-fluoro-2-hydroxybenzoic acid 95%
4-Bromo-5-fluoro-2-hydroxybenzoic acid
Formel:
C₇H₄BrFO₃ Molecular Weight: 235,007 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25459708 CAS-Nummer: 1784366-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...