3-(Difluoromethoxy)-4-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC501941-1G PC501941-250MG PC501941-5G
APOSPC501941-1GEA 46.4 EUR
APOSPC501941-1G APOSPC501941-250MG APOSPC501941-5G
3-(Difluoromethoxy)-4-fluoroaniline
3-(Difluoromethoxy)-4-fluoroaniline
Formel: C₇H₆F₃NO
Molecular Weight: 177,124 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14698444
CAS-Nummer: 83190-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR