4-(Difluoromethoxy)-2-fluoronitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
PC501989-1G
PC501989-25G
PC501989-5G
APOSPC501989-1GEA
55.1
EUR
APOSPC501989-1G
APOSPC501989-25G
APOSPC501989-5G
4-(Difluoromethoxy)-2-fluoronitrobenzene
4-(Difluoromethoxy)-2-fluoronitrobenzene
Formel:
C₇H₄F₃NO₃ Molecular Weight: 207,107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14698452 CAS-Nummer: 1214336-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...