4-Amino-2-Bromo-5-fluorobenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC501990-100MG
PC501990-1G
PC501990-250MG
APOSPC501990-100MGEA
391
EUR
APOSPC501990-100MG
APOSPC501990-1G
APOSPC501990-250MG
4-Amino-2-Bromo-5-fluorobenzonitrile
4-Amino-2-Bromo-5-fluorobenzonitrile
Formel:
C₇H₄BrFN₂ Molecular Weight: 215,022 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD29049281 CAS-Nummer: 1850856-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...