2-(Difluoromethoxy)-4-fluoronitrobenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Difluoromethoxy)-4-fluoro-1-nitrobenzene

PC501991-1G PC501991-250MG PC501991-5G PC501991-10G PC501991-25G
APOSPC501991-1GEA 168 EUR
APOSPC501991-1G APOSPC501991-250MG APOSPC501991-5G APOSPC501991-10G APOSPC501991-25G
2-(Difluoromethoxy)-4-fluoronitrobenzene
2-(Difluoromethoxy)-4-fluoronitrobenzene
Formel: C₇H₄F₃NO₃
Molecular Weight: 207.107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698446
CAS-Nummer: 1214329-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR