2-Bromo-4,6-difluorobenzonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502012-1G
PC502012-250MG
APOSPC502012-1GEA
422
EUR
APOSPC502012-1G
APOSPC502012-250MG
2-Bromo-4,6-difluorobenzonitrile 95%
2-Bromo-4,6-difluorobenzonitrile
Formel:
C₇H₂BrF₂N Molecular Weight: 217,998 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28401849 CAS-Nummer: 1337606-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...