3-Iodo-5-(trifluoromethyl)phenacyl bromide

Lieferant: Apollo Scientific
Gefahr

PC502024-1G PC502024-250MG PC502024-5G
APOSPC502024-1GEA 248 EUR
APOSPC502024-1G APOSPC502024-250MG APOSPC502024-5G
3-Iodo-5-(trifluoromethyl)phenacyl bromide
3-Iodo-5-(trifluoromethyl)phenacyl bromide
Formel: C₉H₅BrF₃IO
Molecular Weight: 392,939 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28060517
CAS-Nummer: 1823897-02-4
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR