3-Bromo-2-(difluoromethoxy)-5-fluoroaniline 95%

Lieferant: Apollo Scientific
Gefahr

PC502026-1G PC502026-250MG
APOSPC502026-1GEA 567 EUR
APOSPC502026-1G APOSPC502026-250MG
3-Bromo-2-(difluoromethoxy)-5-fluoroaniline 95%
3-Bromo-2-(difluoromethoxy)-5-fluoroaniline
Formel: C₇H₅BrF₃NO
Molecular Weight: 256.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12166253
CAS-Nummer: 1096884-90-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR