1-Bromo-1,1,2,2-tetrafluoroheptane

Lieferant: Apollo Scientific
Achtung

PC50203-10G PC50203-1G PC50203-5G
APOSPC50203-10GEA 300 EUR
APOSPC50203-10G APOSPC50203-1G APOSPC50203-5G
1-Bromo-1,1,2,2-tetrafluoroheptane
1-Bromo-1,1,2,2-tetrafluoroheptane
Formel: C₇H₁₁BrF₄
Molecular Weight: 251,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25541898
CAS-Nummer: 1445995-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR