6-(Trifluoromethyl)-1H-benzimidazole

Lieferant: Apollo Scientific

Synonyme: 6-(Trifluoromethyl)-1H-benzo[d]imidazole

PC502047-1G PC502047-5G
APOSPC502047-1GEA 68.6 EUR
APOSPC502047-1G APOSPC502047-5G
6-(Trifluoromethyl)-1H-benzimidazole
6-(Trifluoromethyl)-1H-benzimidazole
Formel: C₈H₅F₃N₂
Molecular Weight: 186.134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027453
CAS-Nummer: 326-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR