3-bromo-4-chloro-5-fluorobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-4-chloro-5-fluorobenzoic acid

PC502059-1G PC502059-250MG PC502059-5G
APOSPC502059-1GEA 150 EUR
APOSPC502059-1G APOSPC502059-250MG APOSPC502059-5G
3-bromo-4-chloro-5-fluorobenzoic acid 95%
3-bromo-4-chloro-5-fluorobenzoic acid
Formel: C₇H₃BrClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21364899
CAS-Nummer: 1357942-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR