3-Bromo-2-(trifluoromethyl)benzyl bromide 95%

Lieferant: Apollo Scientific
Achtung

PC502064-1G PC502064-250MG PC502064-5G
APOSPC502064-1GEA 190 EUR
APOSPC502064-1G APOSPC502064-250MG APOSPC502064-5G
3-Bromo-2-(trifluoromethyl)benzyl bromide 95%
3-Bromo-2-(trifluoromethyl)benzyl bromide
Formel: C₈H₅Br₂F₃
Molecular Weight: 317.929 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28746753
CAS-Nummer: 1428940-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR