3-Bromo-2-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC502065-1G PC502065-250MG PC502065-5G
APOSPC502065-1GEA 0 EUR
APOSPC502065-1G APOSPC502065-250MG APOSPC502065-5G
3-Bromo-2-(trifluoromethyl)benzyl alcohol
3-Bromo-2-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD28746751
CAS-Nummer: 1805950-70-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR