Methyl 3-bromo-4-chloro-5-fluorobenzoate 96%

Lieferant: Apollo Scientific
Achtung

PC502099-1G PC502099-5G PC502099-25G
APOSPC502099-1GEA 43.5 EUR
APOSPC502099-1G APOSPC502099-5G APOSPC502099-25G
Methyl 3-bromo-4-chloro-5-fluorobenzoate 96%
Methyl 3-bromo-4-chloro-5-fluorobenzoate
Formel: C₈H₅BrClFO₂
Molecular Weight: 267.48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846086
CAS-Nummer: 1160574-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR