1,1,1-Trifluorohex-2-ene

Lieferant: Apollo Scientific
Gefahr

PC50212-1G
APOSPC50212-1GEA 50.9 EUR
APOSPC50212-1G
1,1,1-Trifluorohex-2-ene
1,1,1-Trifluorohex-2-ene
Formel: C₆H₉F₃
Molecular Weight: 138,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28136602
CAS-Nummer: 53392-87-3
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR