4-Fluoro-2-nitrobenzyl alcohol
Lieferant: Apollo Scientific
Synonyme:
(4-Fluoro-2-nitrophenyl)methanol
PC502133-5G
APOSPC502133-5GEA
106
EUR
APOSPC502133-5G
4-Fluoro-2-nitrobenzyl alcohol
4-Fluoro-2-nitrobenzyl alcohol
Formel:
C₇H₆FNO₃ Molecular Weight: 171,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368344 CAS-Nummer: 1043416-40-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...