5-Fluoro-2-nitrobenzyl alcohol 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-nitrobenzyl Alcohol

PC502138-1G PC502138-250MG PC502138-5G
APOSPC502138-1GEA 168 EUR
APOSPC502138-1G APOSPC502138-250MG APOSPC502138-5G
5-Fluoro-2-nitrobenzyl alcohol 95+%
5-Fluoro-2-nitrobenzyl alcohol
Formel: C₇H₆FNO₃
Molecular Weight: 171,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202666
CAS-Nummer: 287121-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR