3-Iodo-5-(trifluoromethyl)benzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC502146-1G PC502146-250MG PC502146-5G
APOSPC502146-1GEA 1400 EUR
APOSPC502146-1G APOSPC502146-250MG APOSPC502146-5G
3-Iodo-5-(trifluoromethyl)benzyl bromide
3-Iodo-5-(trifluoromethyl)benzyl bromide
Formel: C₈H₅BrF₃I
Molecular Weight: 364.929 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398193
CAS-Nummer: 1261795-42-9
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR