3-Iodo-5-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC502153-1G PC502153-250MG PC502153-5G
APOSPC502153-1GEA 55.1 EUR
APOSPC502153-1G APOSPC502153-250MG APOSPC502153-5G
3-Iodo-5-(trifluoromethyl)benzoic acid
3-Iodo-5-(trifluoromethyl)benzoic acid
Formel: C₈H₄F₃IO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08448293
CAS-Nummer: 28186-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR