4-Bromo-2-chloro-3-fluoroiodobenzene 95%

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-3-chloro-2-fluoro-4-iodobenzene

PC502154-1G PC502154-250MG PC502154-5G
APOSPC502154-1GEA 200 EUR
APOSPC502154-1G APOSPC502154-250MG APOSPC502154-5G
4-Bromo-2-chloro-3-fluoroiodobenzene 95%
4-Bromo-2-chloro-3-fluoroiodobenzene
Formel: C₆H₂BrClFI
Molecular Weight: 335,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30183258
CAS-Nummer: 1917307-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR