Potassium [3-(trifluoromethyl)benzyl]trifluoroborate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium trifluoro(3-(trifluoromethyl)benzyl)borate
PC502198-1G
PC502198-250MG
PC502198-5G
APOSPC502198-1GEA
333
EUR
APOSPC502198-1G
APOSPC502198-250MG
APOSPC502198-5G
Potassium [3-(trifluoromethyl)benzyl]trifluoroborate 95%
Potassium [3-(trifluoromethyl)benzyl]trifluoroborate
Formel:
C₈H₆BF₆·K Molecular Weight: 266,033 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18712965 CAS-Nummer: 1494466-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...