2,4-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC502199-1G PC502199-250MG
APOSPC502199-1GEA 654 EUR
APOSPC502199-1G APOSPC502199-250MG
2,4-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%
2,4-Difluoro-3-(trifluoromethoxy)benzyl bromide
Formel: C₈H₄BrF₅O
Molecular Weight: 291.013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30723444
CAS-Nummer: 2149591-35-3
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR