5-Bromo-2-fluoro-3-nitrobenzotrifluoride 95%

Lieferant: Apollo Scientific

PC502236-1G PC502236-250MG
APOSPC502236-1GEA 534 EUR
APOSPC502236-1G APOSPC502236-250MG
5-Bromo-2-fluoro-3-nitrobenzotrifluoride 95%
5-Bromo-2-fluoro-3-nitrobenzotrifluoride
Formel: C₇H₂BrF₄NO₂
Molecular Weight: 287.994 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27931581
CAS-Nummer: 889459-12-5
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR