2-Ethynyl-6-fluorobenzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

PC502252-1G PC502252-250MG PC502252-5G
APOSPC502252-1GEA 473 EUR
APOSPC502252-1G APOSPC502252-250MG APOSPC502252-5G
2-Ethynyl-6-fluorobenzaldehyde 98%
2-Ethynyl-6-fluorobenzaldehyde
Formel: C₉H₅FO
Molecular Weight: 148.134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12545737
CAS-Nummer: 137338-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR