2-(4-Amino-2-fluorophenyl)acetonitrile

Lieferant: Apollo Scientific
Achtung

PC50226-1G PC50226-250MG PC50226-5G
APOSPC50226-1GEA 429 EUR
APOSPC50226-1G APOSPC50226-250MG APOSPC50226-5G
2-(4-Amino-2-fluorophenyl)acetonitrile
2-(4-Amino-2-fluorophenyl)acetonitrile
Formel: C₈H₇FN₂
Molecular Weight: 150,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20696905
CAS-Nummer: 180146-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR