4-Bromo-1-(difluoromethoxy)isoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
PC502287-1G
PC502287-250MG
PC502287-5G
APOSPC502287-1GEA
145
EUR
APOSPC502287-1G
APOSPC502287-250MG
APOSPC502287-5G
4-Bromo-1-(difluoromethoxy)isoquinoline
4-Bromo-1-(difluoromethoxy)isoquinoline
Formel:
C₁₀H₆BrF₂NO Molecular Weight: 274,062 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29761120 CAS-Nummer: 2089319-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...