2-Bromo-3’-fluoropropiophenone

Lieferant: Apollo Scientific
Gefahr

PC502288-1G PC502288-250MG PC502288-5G
APOSPC502288-1GEA 36.4 EUR
APOSPC502288-1G APOSPC502288-250MG APOSPC502288-5G
2-Bromo-3’-fluoropropiophenone
2-Bromo-3’-fluoropropiophenone
Formel: C₉H₈BrFO
Molecular Weight: 231,062 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09751763
CAS-Nummer: 92821-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR