4-Bromo-5-(difluoromethoxy)-2-fluoronitrobenzene

Lieferant: Apollo Scientific
Achtung

PC502333-1G PC502333-250MG PC502333-5G
APOSPC502333-1GEA 596 EUR
APOSPC502333-1G APOSPC502333-250MG APOSPC502333-5G
4-Bromo-5-(difluoromethoxy)-2-fluoronitrobenzene
Formel: C₇H₃BrF₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22580935
CAS-Nummer: 1417566-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR