2-Bromo-2',5'-difluoropropiophenone
Lieferant: Apollo Scientific
|
Gefahr
|
PC502334-1G
PC502334-250MG
PC502334-5G
APOSPC502334-1GEA
58
EUR
APOSPC502334-1G
APOSPC502334-250MG
APOSPC502334-5G
2-Bromo-2',5'-difluoropropiophenone
2-Bromo-2',5'-difluoropropiophenone
Formel:
C₉H₇BrF₂O Molecular Weight: 249.052 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11182341 CAS-Nummer: 186036-04-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...