3,4-Difluoro-2-(difluoromethoxy)aniline

Lieferant: Apollo Scientific
Achtung

PC502340-1G PC502340-250MG
APOSPC502340-1GEA 474 EUR
APOSPC502340-1G APOSPC502340-250MG
3,4-Difluoro-2-(difluoromethoxy)aniline
3,4-Difluoro-2-(difluoromethoxy)aniline
Formel: C₇H₅F₄NO
Molecular Weight: 195.114 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20723017
CAS-Nummer: 206546-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR