3-Fluoro-4-methoxy-5-nitrobenzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502390-1G
PC502390-250MG
APOSPC502390-1GEA
521
EUR
APOSPC502390-1G
APOSPC502390-250MG
3-Fluoro-4-methoxy-5-nitrobenzoic acid 95%
3-Fluoro-4-methoxy-5-nitrobenzoic acid
Formel:
C₈H₆FNO₅ Molecular Weight: 215,135 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12173014 CAS-Nummer: 577-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...