3-Bromo-2,5,6-trifluorobenzoic acid 98%

Lieferant: Apollo Scientific

PC502419-10G PC502419-1G PC502419-5G
APOSPC502419-10GEA 708 EUR
APOSPC502419-10G APOSPC502419-1G APOSPC502419-5G
3-Bromo-2,5,6-trifluorobenzoic acid 98%
3-Bromo-2,5,6-trifluorobenzoic acid
Formel: C₇H₂BrF₃O₂
Molecular Weight: 254.989 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25956267
CAS-Nummer: 118829-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR