2-Bromo-3-fluoro-6-methoxyaniline 95%

Lieferant: Apollo Scientific
Achtung

PC502424-1G PC502424-250MG PC502424-5G
APOSPC502424-1GEA 575 EUR
APOSPC502424-1G APOSPC502424-250MG APOSPC502424-5G
2-Bromo-3-fluoro-6-methoxyaniline 95%
2-Bromo-3-fluoro-6-methoxyaniline
Formel: C₇H₇BrFNO
Molecular Weight: 220,039 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29481964
CAS-Nummer: 1698500-38-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR